Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N,N',N'-Tetraacetylethylenediamine 90%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

34459-0500 34459-2500
TS34459-0500EA 65 USD InStock
TS34459-0500 TS34459-2500
N,N,N',N'-Tetraacetylethylenediamine 90%
N,N,N',N'-Tetraacetylethylenediamine


Formula: C₁₀H₁₆N₂O₄
Boiling Pt: 140 °C (2hPa)
Melting Pt: 149…154 °C
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00014967
CAS Number: 10543-57-4
Order Now
Specification Test Results
Appearance Off-white to beige Granular powder
Infrared spectrum Conforms
Assay ≥88 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty