Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Pyrrolidinecarbonyl chloride 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

41865-0050 41865-0250
TS41865-0050EA 67 USD InStock
TS41865-0050 TS41865-0250
1-Pyrrolidinecarbonyl chloride 97%
1-Pyrrolidinecarbonyl chloride


Formula: C₅H₈ClNO
Boiling Pt: 75 °C (1 mmHg)
Density: 1.209 g/cm³
Flash Pt: 98 °C
Storage Temperature: Freezer
MDL Number: MFCD00051321
CAS Number: 1192-63-8
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4910 to 1.4970 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty