Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-1-hexene 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

36470-0010 36470-0050 36470-0250
TS36470-0010EA 50 USD InStock
TS36470-0010 TS36470-0050 TS36470-0250
6-Bromo-1-hexene 97%
6-Bromo-1-hexene


Formula: C₆H₁₁Br
Boiling Pt: 47…51 °C (16 mmHg)
Density: 1.217 g/cm³
Flash Pt: 52 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000269
CAS Number: 2695-47-8
EINECS: 220-267-9
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4640 to 1.4670 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty