Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3-Dimethoxypropiononitrile 90%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

44626-0250 44626-1000
TS44626-0250EA 159 USD InStock
TS44626-0250 TS44626-1000
3,3-Dimethoxypropiononitrile 90%
3,3-Dimethoxypropiononitrile


Formula: C₅H₉NO₂
Boiling Pt: 195 °C (760 mmHg)
Density: 1.026 g/cm³
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00044797
CAS Number: 57597-62-3
Order Now
Specification Test Results
Appearance Colorless to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4120 to 1.4160 (20°C, 589 nm)
GC ≥88 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty