2-(Methylsulphonyl)phenol 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

45575-0010 45575-0050
TS45575-0010EA 109 USD
TS45575-0010 TS45575-0050
2-(Methylsulphonyl)phenol 98%
2-(Methylsulphonyl)phenol
Formula: C₇H₈O₃S
Melting Pt: 88…90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00127516
CAS Number: 27489-33-4

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 87 to 92 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR