Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-1H-pyrazole 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

45992-0050 45992-0250
TS45992-0050EA 71 USD InStock
TS45992-0050 TS45992-0250
4-Chloro-1H-pyrazole 98%
4-Chloro-1H-pyrazole


Formula: C₃H₃ClN₂
MW: 102.5200 g/mol
Melting Pt: 71…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00159634
CAS Number: 15878-00-9
Order Now
Specification Test Results
Appearance White to brown Powder or crystalline powder or solid
Infrared spectrum Conforms
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty