Imidazo[1,2-a]pyridine-7-carboxylic acid hydrochloride 97%
Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
|
Warning
|
45336-0010
45336-0050
TS45336-0010EA
113
USD
TS45336-0010
TS45336-0050
Imidazo[1,2-a]pyridine-7-carboxylic acid hydrochloride 97%
Imidazo[1,2-a]pyridine-7-carboxylic acid hydrochloride
Formula:
C₈H₆N₂O₂·ClH MW: 198.61 g/mol Melting Pt: 295…298 °C Storage Temperature: Ambient |
MDL Number:
MFCD09954929 CAS Number: 1423031-35-9 |
Specification Test Results
Appearance | White to yellow Powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥96.0 to ≤104.0 % |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...