Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Chloromethyl)benzimidazole 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

10939-0050 10939-0250
TS10939-0050EA 43 USD InStock
TS10939-0050 TS10939-0250
2-(Chloromethyl)benzimidazole 95%
2-(Chloromethyl)benzimidazole


Formula: C₈H₇ClN₂
Melting Pt: 145…150 °C
MDL Number: MFCD00005602
CAS Number: 4857-04-9
EINECS: 225-452-8
Order Now
Specification Test Results
Appearance White to yellow to brown Powder
Infrared spectrum Conforms
Melting point 143 to 150 °C
HPLC ≥94 %
Titration with HClO4 94 to 106 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty