Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitropyrocatechol 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12860-0050 12860-0100 12860-0500
TS12860-0050EA 57 USD InStock
TS12860-0050 TS12860-0100 TS12860-0500
4-Nitropyrocatechol 97%
4-Nitropyrocatechol


Formula: C₆H₅NO₄
Melting Pt: 173…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00007242
CAS Number: 3316-09-4
EINECS: 222-009-0
Order Now
Specification Test Results
Appearance Yellow to yellow-green to yellow brown Crystalline powder
Infrared spectrum Conforms
Melting point 173 to 177 °C
Titration with NaOH ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty