Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Decanoyl chloride 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

16119-1000 16119-5000
TS16119-1000EA 45 USD InStock
TS16119-1000 TS16119-5000
Decanoyl chloride 98+%
Decanoyl chloride


Formula: C₁₀H₁₉ClO
Boiling Pt: 233 °C (1013 hPa)
Melting Pt: –34.5 °C
Density: 0.919 g/cm³
Flash Pt: 110 °C
MDL Number: MFCD00000771
CAS Number: 112-13-0
EINECS: 203-938-0
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4430 (20°C, 589 nm)
GC ≥98.0 %
Color scale ≤50 APHA
Carboxylic acid ≤2 % (as Decanoic acid)
Titration RCOCl ≥98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty