Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl myristate 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

16501-0025 16501-1000 16501-5000
TS16501-0025EA 31 USD InStock
TS16501-0025 TS16501-1000 TS16501-5000
Methyl myristate 99%
Methyl myristate


Formula: C₁₅H₃₀O₂
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 18.4 °C
Density: 0.863 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008983
CAS Number: 124-10-7
EINECS: 204-680-1
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4340 to 1.4380 (20 °C, 589 nm)
Density (20/4°C) 0.853 to 0.873 g/ml
GC ≥98.5 %
Acid value ≤1 mg KOH/g

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty