Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium (+/-)-10-camphorsulfonate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

18912-0050 18912-5000
TS18912-0050EA 24 USD InStock
TS18912-0050 TS18912-5000
Sodium (+/-)-10-camphorsulfonate 98%
Sodium (+/-)-10-camphorsulfonate


Formula: C₁₀H₁₆O₄S
Melting Pt: 286…288 °C
Storage Temperature: Ambient
MDL Number: MFCD00135623
CAS Number: 34850-66-3
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty