Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,4,5,6,6-Hexachloro-2,4-cyclohexadien-1-one 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

24395-0050 24395-0250
TS24395-0050EA 121 USD InStock
TS24395-0050 TS24395-0250
2,3,4,5,6,6-Hexachloro-2,4-cyclohexadien-1-one 99%
2,3,4,5,6,6-Hexachloro-2,4-cyclohexadien-1-one


Formula: C₆Cl₆O
Melting Pt: 46…49 °C
Storage Temperature: Ambient
MDL Number: MFCD00019431
CAS Number: 21306-21-8
Order Now
Specification Test Results
Appearance Yellow to brown Crystalline solid
Infrared spectrum Conforms
Melting point 46 to 52 °C
HPLC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty