Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-1-butanol 85% balance THF and HCl

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

35631-0100 35631-1000 35631-5000
TS35631-0100EA 34 USD InStock
TS35631-0100 TS35631-1000 TS35631-5000
4-Chloro-1-butanol 85% balance THF and HCl
4-Chloro-1-butanol


Formula: C₄H₉ClO
Boiling Pt: 58…60 °C (3 mmHg)
Density: 1.089 g/cm³
Flash Pt: 36 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002967
CAS Number: 928-51-8
EINECS: 213-175-5
UN: 2924
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow to light pink Liquid
Infrared spectrum Conforms
Refractive index 1.4510 to 1.4630 (20 °C, 589 nm)
GC ≥80 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty