Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl linoleate 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

42813-0050 42813-0250 42813-1000
TS42813-0050EA 86 USD InStock
TS42813-0050 TS42813-0250 TS42813-1000
Methyl linoleate 99%
Methyl linoleate


Formula: C₁₉H₃₄O₂
MW: 294.48 g/mol
Boiling Pt: 340
Melting Pt: -35
Density: 0.89 g/cm³
Flash Pt: >200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009534
CAS Number: 112-63-0
EINECS: 203-993-0
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4610 to 1.4630 (20 °C, 589 nm)
GC ≥98.5 %
Specific optical rotation -85° to -95° (20°C, 589 nm) (c=1, CHCl3)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty