3,4-Dimethoxyaniline 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

10472-0100 10472-0250
TS10472-0100EA 49 USD
TS10472-0100 TS10472-0250
3,4-Dimethoxyaniline 98%
3,4-Dimethoxyaniline
Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
Boiling Pt: 296
Melting Pt: 86…88 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008394
CAS Number: 6315-89-5
EINECS: 228-647-6
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Brown Crystalline powder
Infrared spectrum Conforms
Melting point 86 to 88 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR