Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzo[ghi]perylene 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

10555-0050 10555-0250 10555-1000
TS10555-0050EA 43 USD InStock
TS10555-0050 TS10555-0250 TS10555-1000
Benzo[ghi]perylene 98+%
Benzo[ghi]perylene


Formula: C₂₂H₁₂
Boiling Pt: >500 °C (760 mmHg)
Melting Pt: 276…280 °C
Storage Temperature: Ambient
MDL Number: MFCD00004135
CAS Number: 191-24-2
EINECS: 205-883-8
Order Now
Specification Test Results
Appearance Yellow to orange Crystalline powder or crystals or needles
Infrared spectrum Conforms
Melting point 276 to 280 °C
GC-MS ≥98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty