3,5-Dimethoxyaniline 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

11536-0050 11536-0250 11536-1000
TS11536-0050EA 47 USD
TS11536-0050 TS11536-0250 TS11536-1000
3,5-Dimethoxyaniline 98%
3,5-Dimethoxyaniline
Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
Boiling Pt: 303
Melting Pt: 53…55 °C
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00008392
CAS Number: 10272-07-8
EINECS: 233-616-5

Order Now

Specification Test Results

Appearance White to beige to grey to light brown Powder or crystals or needles
Infrared spectrum Conforms
Melting point 54 to 57 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR