Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxybenzophenone 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

14692-0010 14692-0100 14692-0250
TS14692-0010EA 25 USD InStock
TS14692-0010 TS14692-0100 TS14692-0250
2-Hydroxybenzophenone 99%
2-Hydroxybenzophenone


Formula: C₁₃H₁₀O₂
Boiling Pt: 171…173 °C (12 mmHg)
Melting Pt: 37…40 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00002216
CAS Number: 117-99-7
EINECS: 204-226-2
Order Now
Specification Test Results
Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 37 to 40 °C
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty