1-tert-Butyl-4-ethynylbenzene 96%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

38085-0050 38085-0250
TS38085-0050EA 71 USD
TS38085-0050 TS38085-0250
1-tert-Butyl-4-ethynylbenzene 96%
1-tert-Butyl-4-ethynylbenzene
Formula: C₁₂H₁₄
MW: 158.24
Boiling Pt: 130 °C (40 mmHg)
Flash Pt: 82.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00190197
CAS Number: 772-38-3
UN: 3082
ADR: 9 III

Order Now

Specification Test Results

Appearance Clear colorless to yellow to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5270 to 1.5310 (20°C, 589 nm)
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR