Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl piperazine-1-carboxylate 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

11887-0250 11887-1000 11887-2500
TS11887-0250EA 35 USD InStock
TS11887-0250 TS11887-1000 TS11887-2500
Ethyl piperazine-1-carboxylate 99%
Ethyl piperazine-1-carboxylate


Formula: C₇H₁₄N₂O₂
Boiling Pt: 273 °C (760 mmHg)
Density: 1.08 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005964
CAS Number: 120-43-4
EINECS: 204-395-2
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4760 to 1.4790 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty