Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hexylthiophene 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

36738-0050 36738-0250
TS36738-0050EA 105 USD InStock
TS36738-0050 TS36738-0250
3-Hexylthiophene 98%
3-Hexylthiophene


Formula: C₁₀H₁₆S
Boiling Pt: 65 °C (.4 mmHg)
Density: 0.936 g/cm³
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00143181
CAS Number: 1693-86-3
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4930 to 1.4990 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty