Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-fluoropyridine 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

43742-0010 43742-0050
TS43742-0010EA 48 USD InStock
TS43742-0010 TS43742-0050
3-Bromo-5-fluoropyridine 97%
3-Bromo-5-fluoropyridine


Formula: C₅H₃BrFN
Boiling Pt: 78 °C (11Torr)
Melting Pt: 24…28 °C
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD04112555
CAS Number: 407-20-5
Order Now
Specification Test Results
Appearance Colorless to white Liquid or low melting solid
Infrared spectrum Conforms
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty