α,3,4-Trichlorotoluene 98+%
Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
|
Danger
|
27905-0250
27905-1000
27905-5000
TS27905-0250EA
30
USD
TS27905-0250
TS27905-1000
TS27905-5000
α,3,4-Trichlorotoluene 98+%
α,3,4-Trichlorotoluene
Formula:
C₇H₅Cl₃ MW: 195.47 g/mol Boiling Pt: 241 Melting Pt: 37.5 Density: 1.41 g/cm³ Flash Pt: 155 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000906 CAS Number: 102-47-6 EINECS: 203-033-0 UN: 3265 ADR: 8 III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5756 to 1.5776 (20°C, 589 nm) |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...