4-(Trifluoromethoxy)phenol 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

30604-0010 30604-0050
TS30604-0010EA 28 USD
TS30604-0010 TS30604-0050
4-(Trifluoromethoxy)phenol 98%
4-(Trifluoromethoxy)phenol
Formula: C₇H₅F₃O₂
MW: 178.11 g/mol
Boiling Pt: 180
Melting Pt: 17…18 °C
Density: 1.37 g/cm³
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00040988
CAS Number: 828-27-3
EINECS: 212-583-0
UN: 2810
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4490 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR