4-Bromo-2-fluorobenzoyl chloride 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

34746-0050 34746-0250
TS34746-0050EA 47 USD
TS34746-0050 TS34746-0250
4-Bromo-2-fluorobenzoyl chloride 99%
4-Bromo-2-fluorobenzoyl chloride
Formula: C₇H₃BrClFO
MW: 237.46 g/mol
Boiling Pt: 86…88 °C (15 mmHg)
Melting Pt: 24…24.5 °C
MDL Number: MFCD03411584
CAS Number: 151982-51-3
UN: 3261
ADR: 8 II