2-Bromo-5-nitrothiophene 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

45115-0010 45115-0050 45115-0250
TS45115-0010EA 44 USD
TS45115-0010 TS45115-0050 TS45115-0250
2-Bromo-5-nitrothiophene 97%
2-Bromo-5-nitrothiophene
Formula: C₄H₂BrNO₂S
MW: 208.04
Boiling Pt: 235…237 °C (1013 hPa)
Melting Pt: 44…48 °C
Storage Temperature: Ambient
MDL Number: MFCD00022493
CAS Number: 13195-50-1