Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl p-Tolyl sulfone 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR1358-100G OR1358-5G
77404-672EA 28.52 USD InStock
77404-672 77404-674
Methyl p-Tolyl sulfone 98%
Methyl p-Tolyl sulfone


Formula: C₈H₁₀O₂S
Melting Pt: 85…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00014742
CAS Number: 3185-99-7
EINECS: 221-682-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty