Anti-BCL-2 Oncoprotein Mouse Monoclonal Antibody [clone: 100/D5]
Supplier: Diagnostic Biosystems
MOB130-01 MOB130-05 MOB130
76633-358EA 108.26 USD InStock
76633-358 76633-360 76633-362
Anti-BCL-2 Oncoprotein Mouse Monoclonal Antibody [clone: 100/D5]
Antibodies
The BCL-2 product is considered to act as an inhibitor of apoptosis. BCL-2 expression is inhibited in germinal centers where apoptosis forms part of the B cell production pathway. Reactive follicles show no staining for BCL-2, whereas the cells in neoplastic follicles exhibit membrane staining. BCL-2 α oncoprotein has been shown to inhibit apoptosis.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty