Anti-MGMT Mouse Monoclonal Antibody [clone: MT3.1]
Supplier: Diagnostic Biosystems
MOB423-01 MOB423-05 MOB423-RUO
76633-862EA 140.11 USD
76633-862 76633-864 76633-866
Anti-MGMT Mouse Monoclonal Antibody [clone: MT3.1]
Antibodies
O(6)-methylguanine-DNA-methyltransferase (MGMT) is a 22 kDa human DNA repair protein that removes O(6)-alkylguanine DNA adducts. MGMT acts as a suicide acceptor protein repairing the DNA by accepting alkyl groups and consequently inactivating itself. This antibody stains all mantle zone lymphocytes and 50% of germinal center lymphocytes. Basaloid epithelial cells of tonsil squamous mucosa are also stained positive with this antibody.