Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Loreclezole 97%

Supplier: AMBEED, INC
Warning

A1001459-10MG A1001459-1MG A1001459-5MG
76645-884EA 48 USD InStock
76645-884 76645-886 76645-888
Loreclezole 97%
Loreclezole
Precaución: for Research Use Only


Formula: C₁₀H₆Cl₃N₃
MW: 274.53 g/mol
MDL Number: MFCD00865331
CAS Number: 117857-45-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty