Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,2,4]triazolo[1,5-a]pyridine 95% (contains 30-35% Pinacone)

Supplier: AMBEED, INC
Warning

A1003432-100MG A1003432-250MG
76646-138EA 59 USD InStock
76646-138 77619-506
7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,2,4]triazolo[1,5-a]pyridine 95% (contains 30-35% Pinacone)
7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,2,4]triazolo[1,5-a]pyridine
Precaución: for Research Use Only


Formula: C₁₂H₁₆BN₃O₂
MDL Number: MFCD13181995
CAS Number: 1210048-18-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty