Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 3,5-difluoropicolinate 95+%

Supplier: AMBEED, INC
Warning

A334845-100MG A334845-250MG
76648-360EA 15 USD InStock
76648-360 76648-362
tert-Butyl 3,5-difluoropicolinate 95+%
tert-Butyl 3,5-difluoropicolinate
Precaución: for Research Use Only


Formula: C₁₀H₁₁F₂NO₂
CAS Number: 1402742-26-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty