Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chromane-7-carbaldehyde 95%

Supplier: AMBEED, INC
Warning

A455916-100MG A455916-250MG A455916-1G
76649-092EA 157 USD InStock
76649-092 76649-094 77748-924
Chromane-7-carbaldehyde 95%
Chromane-7-carbaldehyde
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₂
MDL Number: MFCD24678164
CAS Number: 124362-47-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty