Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,7-Dimethoxybenzo[d]isothiazol-3(2H)-one 1,1-dioxide 95%

Supplier: AMBEED, INC
Warning

A482447-100MG A482447-1G A482447-250MG
76649-280EA 19 USD InStock
76649-280 76649-282 76649-284
5,7-Dimethoxybenzo[d]isothiazol-3(2H)-one 1,1-dioxide 95%
5,7-Dimethoxybenzo[d]isothiazol-3(2H)-one 1,1-dioxide
Precaución: for Research Use Only


Formula: C₉H₉NO₅S
MDL Number: MFCD24369284
CAS Number: 64404-51-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty