Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-4,5,6,7-tetrahydropyrazolo[1,5-a]pyridine 95%

Supplier: AMBEED, INC
Warning

A512033-100MG A512033-250MG
76649-516EA 41 USD InStock
76649-516 76649-518
3-Chloro-4,5,6,7-tetrahydropyrazolo[1,5-a]pyridine 95%
3-Chloro-4,5,6,7-tetrahydropyrazolo[1,5-a]pyridine
Precaución: for Research Use Only


Formula: C₇H₉ClN₂
MDL Number: MFCD30528493
CAS Number: 2138386-72-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty