Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl (S)-3-(cyanomethyl)piperazine-1-carboxylate 98%

Supplier: AMBEED, INC
Warning

A561849-100MG A561849-250MG A561849-50MG
76649-806EA 47 USD InStock
76649-806 76649-808 76649-810
Benzyl (S)-3-(cyanomethyl)piperazine-1-carboxylate 98%
Benzyl (S)-3-(cyanomethyl)piperazine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₄H₁₇N₃O₂
MDL Number: MFCD09263273
CAS Number: 1242267-82-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty