Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2,6-Dioxopiperidin-3-yl)-5-hydroxyisoindoline-1,3-dione 95%

Supplier: AMBEED, INC
Warning

A571532-100MG A571532-10G A571532-1G A571532-250MG A571532-25G A571532-5G
76649-874EA 8 USD InStock
76649-874 76649-876 76649-878 76649-880 76649-882 76649-884
2-(2,6-Dioxopiperidin-3-yl)-5-hydroxyisoindoline-1,3-dione 95%
2-(2,6-Dioxopiperidin-3-yl)-5-hydroxyisoindoline-1,3-dione
Precaución: for Research Use Only


Formula: C₁₃H₁₀N₂O₅
MDL Number: MFCD24386541
CAS Number: 64567-60-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty