Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-((7-Nitrobenzo[c][1,2,5]oxadiazol-4-yl)thio)pyridine 1-oxide 99%

Supplier: AMBEED, INC
Warning

A608208-100MG A608208-10MG A608208-1MG A608208-25MG A608208-50MG A608208-5MG
76650-146EA 134 USD InStock
76650-146 76650-148 76650-150 76650-152 76650-154 76650-156
2-((7-Nitrobenzo[c][1,2,5]oxadiazol-4-yl)thio)pyridine 1-oxide 99%
2-((7-Nitrobenzo[c][1,2,5]oxadiazol-4-yl)thio)pyridine 1-oxide
Precaución: for Research Use Only


Formula: C₁₁H₆N₄O₄S
MDL Number: MFCD30742994
CAS Number: 113104-25-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty