Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Di-tert-butyl (2-aminopropane-1,3-diyl)dicarbamate 97%

Supplier: AMBEED, INC
Warning

A629759-100MG A629759-250MG
76650-356EA 43 USD InStock
76650-356 76650-358
Di-tert-butyl (2-aminopropane-1,3-diyl)dicarbamate 97%
Di-tert-butyl (2-aminopropane-1,3-diyl)dicarbamate
Precaución: for Research Use Only


Formula: C₁₃H₂₇N₃O₄
MDL Number: MFCD24465597
CAS Number: 149876-86-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty