Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl 3-oxocyclopentanecarboxylate 95%

Supplier: AMBEED, INC
Warning

A668413-100MG A668413-1G A668413-250MG
76650-792EA 19 USD InStock
76650-792 76650-794 76650-796
Benzyl 3-oxocyclopentanecarboxylate 95%
Benzyl 3-oxocyclopentanecarboxylate
Precaución: for Research Use Only


Formula: C₁₃H₁₄O₃
MDL Number: MFCD16036435
CAS Number: 130761-99-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty