Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Octyl itaconate 98%

Supplier: AMBEED, INC
Warning

A717516-1MG A717516-100MG A717516-5G
76651-490EA 10 USD InStock
76651-490 77622-550 77622-552
4-Octyl itaconate 98%
4-Octyl itaconate
Precaución: for Research Use Only


Formula: C₁₃H₂₂O₄
MDL Number: MFCD31706409
CAS Number: 3133-16-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty