Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pentanoate 95%

Supplier: AMBEED, INC
Warning

A739910-100MG A739910-1G A739910-250MG
76652-078EA 46 USD InStock
76652-078 76652-080 76652-082
Methyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pentanoate 95%
Methyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pentanoate
Precaución: for Research Use Only


Formula: C₁₂H₂₃BO₄
MDL Number: MFCD27936503
CAS Number: 1985620-97-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty