Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Palladium on calcium carbonate 5% Pd, moistened with water, unreduced

Supplier: AMBEED, INC
Danger

A752074-1G A752074-25G A752074-5G
76652-390EA 33 USD InStock
76652-390 76652-392 76652-394
Palladium on calcium carbonate 5% Pd, moistened with water, unreduced
Palladium on calcium carbonate
Precaución: for Research Use Only


Formula: CCaO₃Pd
MDL Number: MFCD03427452
CAS Number: 1257650-74-0
UN: 3288
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty