Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Iodo-2-methylbutane 95% stabilized

Supplier: AMBEED, INC
Danger

Stabilization: Stabilized with Copper chip

A752179-1G A752179-250MG
76652-408EA 24 USD InStock
76652-408 76652-410
1-Iodo-2-methylbutane 95% stabilized
1-Iodo-2-methylbutane
Precaución: for Research Use Only


Formula: C₅H₁₁I
MDL Number: MFCD01321177
CAS Number: 616-14-8
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty