Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Methanesulfonylmethyl)aniline 95%

Supplier: AMBEED, INC
Warning

A789020-100MG A789020-1G A789020-250MG A789020-5G
76653-110EA 40 USD InStock
76653-110 76653-112 76653-114 76653-116
2-(Methanesulfonylmethyl)aniline 95%
2-(Methanesulfonylmethyl)aniline
Precaución: for Research Use Only


Formula: C₈H₁₁NO₂S
MDL Number: MFCD12823721
CAS Number: 25195-69-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty