Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-1-(4-(4Methylthiazol-5-yl)phenyl)ethan-1-amine HCl 95%

Supplier: AMBEED, INC
Warning

A820540-100MG A820540-250MG
76653-816EA 14 USD InStock
76653-816 76653-818
(S)-1-(4-(4Methylthiazol-5-yl)phenyl)ethan-1-amine HCl 95%
(S)-1-(4-(4Methylthiazol-5-yl)phenyl)ethan-1-amine HCl
Precaución: For research use only.


Formula: C₁₂H₁₅ClN₂S
MDL Number: MFCD30741910
CAS Number: 1948273-01-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty