Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-3-pentylimidazolium bromide 98% (by HPLC)

Supplier: AMBEED, INC
Warning

A862121-100G A862121-1G A862121-25G A862121-5G
76654-550EA 120 USD InStock
76654-550 76654-552 76654-554 76654-556
1-Methyl-3-pentylimidazolium bromide 98% (by HPLC)
1-Methyl-3-pentylimidazolium bromide
Precaución: for Research Use Only


Formula: C₉H₁₇BrN₂
MDL Number: MFCD19382537
CAS Number: 343851-31-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty