Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)-4-methylpiperazine 95%

Supplier: AMBEED, INC
Warning

A883875-1G A883875-250MG A883875-5G
76654-850EA 67 USD InStock
76654-850 76654-852 76654-854
1-(2-Chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)-4-methylpiperazine 95%
1-(2-Chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)-4-methylpiperazine
Precaución: for Research Use Only


Formula: C₁₈H₂₈BClN₂O₂
MDL Number: MFCD20441909
CAS Number: 2096340-35-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty