Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Aminopentanoic acid tert-butyl ester 95%

Supplier: AMBEED, INC
Warning

A918026-250MG A918026-1G A918026-100MG
76655-360EA 61 USD InStock
76655-360 77569-638 77623-140
5-Aminopentanoic acid tert-butyl ester 95%
5-Aminopentanoic acid tert-butyl ester
Precaución: For research use only.


Formula: C₉H₁₉NO₂
MDL Number: MFCD08275839
CAS Number: 63984-03-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty